2-Amino-3-methylphenylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Carboxymethyl)-6-methylaniline, 2-Amino-3-(carboxymethyl)toluene

OR18018-1G OR18018-250MG
APOSOR18018-1GEA 2970 EUR
APOSOR18018-1G APOSOR18018-250MG
2-Amino-3-methylphenylacetic acid
2-Amino-3-methylphenylacetic acid
Formel: C₉H₁₁NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16606085
CAS-Nummer: 1334405-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR