4-Acetyl-2-chlorobenzoic acid

Lieferant: Apollo Scientific
Achtung

OR17681-1G OR17681-5G OR17681-25G
APOSOR17681-1GEA 132 EUR
APOSOR17681-1G APOSOR17681-5G APOSOR17681-25G
4-Acetyl-2-chlorobenzoic acid
4-Acetyl-2-chlorobenzoic acid
Formel: C₉H₇ClO₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD20639602
CAS-Nummer: 115382-35-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR