4-Amino-3-hydroxybenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR17680-1G
OR17680-250MG
APOSOR17680-1GEA
319
EUR
APOSOR17680-1G
APOSOR17680-250MG
4-Amino-3-hydroxybenzonitrile
4-Amino-3-hydroxybenzonitrile
Formel:
C₇H₆N₂O Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD06657033 CAS-Nummer: 55586-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...