4-Amino-2-bromo-6-methylphenol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromo-4-hydroxy-5-methylaniline, 5-Amino-3-bromo-2-hydroxytoluene, 4-Amino-6-bromo-o-cresol, 5-Bromo-4-hydroxy-m-toluidine

OR17625-1G OR17625-250MG OR17625-5G
APOSOR17625-1GEA 647 EUR
APOSOR17625-1G APOSOR17625-250MG APOSOR17625-5G
4-Amino-2-bromo-6-methylphenol 95%
4-Amino-2-bromo-6-methylphenol
Formel: C₇H₈BrNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD12402419
CAS-Nummer: 848676-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR