1-(2-Aminoethyl)-1H-imidazole dihydrobromide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(1H-Imidazol-1-yl)ethylamine dihydrobromide
OR12977-100MG
OR12977-250MG
APOSOR12977-100MGEA
42.2
EUR
APOSOR12977-100MG
APOSOR12977-250MG
1-(2-Aminoethyl)-1H-imidazole dihydrobromide
1-(2-Aminoethyl)-1H-imidazole dihydrobromide
Formel:
C₅H₉N₃·2BrH Molekulargewicht: 272.969 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04117725 CAS-Nummer: 167298-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...