5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-borono-4-methoxybenzoate, 5-(Ethoxycarbonyl)-2-methoxyphenylboronic acid, 2-Borono-4-(ethoxycarbonyl)anisole

OR11693-1G OR11693-250MG OR11693-5G
APOSOR11693-1GEA 177 EUR
APOSOR11693-1G APOSOR11693-250MG APOSOR11693-5G
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid 95%
5-(Ethoxycarbonyl)-2-methoxybenzeneboronic acid
Formel: C₁₀H₁₃BO₅
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09800892
CAS-Nummer: 957062-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR