4-(4-Methylphenoxy)-1H-indazol-3-amine

Lieferant: Apollo Scientific

OR110880-1G OR110880-250MG OR110880-5G
APOSOR110880-1GEA 348 EUR
APOSOR110880-1G APOSOR110880-250MG APOSOR110880-5G
4-(4-Methylphenoxy)-1H-indazol-3-amine
4-(4-Methylphenoxy)-1H-indazol-3-amine
Formel: C₁₄H₁₃N₃O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD26939170
CAS-Nummer: 871708-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR