N-Methyl-4-pentylbenzamide

Lieferant: Apollo Scientific

OR110452-1G OR110452-250MG OR110452-5G
APOSOR110452-1GEA 186 EUR
APOSOR110452-1G APOSOR110452-250MG APOSOR110452-5G
N-Methyl-4-pentylbenzamide
N-Methyl-4-pentylbenzamide
Formel: C₁₃H₁₉NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03551083
CAS-Nummer: 401587-39-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR