Methyl 6-amino-5-chloronicotinate
Lieferant: Apollo Scientific
|
Gefahr
|
OR110263-1G
OR110263-5G
APOSOR110263-1GEA
132
EUR
APOSOR110263-1G
APOSOR110263-5G
Methyl 6-amino-5-chloronicotinate
Methyl 6-amino-5-chloronicotinate
Formel:
C₇H₇ClN₂O₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD12964045 CAS-Nummer: 856211-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...