4-Sulphamoylbutanoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR10407-1G
OR10407-5G
APOSOR10407-1GEA
284
EUR
APOSOR10407-1G
APOSOR10407-5G
4-Sulphamoylbutanoic acid 97%
4-Sulphamoylbutanoic acid
Formel:
C₄H₉NO₄S Schmelzpunkt: 90 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01075007 CAS-Nummer: 175476-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...