Dicyclopropylmethylamine hydrochloride 97%

Lieferant: Apollo Scientific
Achtung

OR0939-1G OR0939-10G OR0939-5G
APOSOR0939-1GEA 102 EUR
APOSOR0939-1G APOSOR0939-10G APOSOR0939-5G
Dicyclopropylmethylamine hydrochloride 97%
Dicyclopropylmethylamine hydrochloride
Formel: C₇H₁₃N·ClH
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD08059568
CAS-Nummer: 51043-72-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR