5-Cyano-6-cyclohexyl-2-thiouracil 97%
Lieferant: Apollo Scientific
OR0307-1G
OR0307-5G
APOSOR0307-1GEA
72.5
EUR
APOSOR0307-1G
APOSOR0307-5G
5-Cyano-6-cyclohexyl-2-thiouracil 97%
5-Cyano-6-cyclohexyl-2-thiouracil
Formel:
C₁₁H₁₃N₃OS Schmelzpunkt: >300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01312949 CAS-Nummer: 290313-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...