1-(Naphth-1-yl)piperazine dihydrochloride 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
1-(Piperazin-1-yl)naphthalene dihydrochloride
OR0079-1G
OR0079-250MG
APOSOR0079-1GEA
207
EUR
APOSOR0079-1G
APOSOR0079-250MG
1-(Naphth-1-yl)piperazine dihydrochloride 97%
1-(Naphth-1-yl)piperazine dihydrochloride
Formel:
C₁₄H₁₆N₂·2ClH Schmelzpunkt: >260 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03540959 CAS-Nummer: 1188264-04-1 UN-Nummer: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...