Fmoc-N-amido-PEG7-acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(9H-Fluoren-9-yl)-3-oxo-2,7,10,13,16,19,22,25-octaoxa-4-azaoctacosan-28-oic acid, Fmoc-N-PEG7-acid
BIPG1401-500MG
BIPG1401-100MG
APOSBIPG1401-500MGEA
415
EUR
APOSBIPG1401-500MG
APOSBIPG1401-100MG
Fmoc-N-amido-PEG7-acid
Fmoc-N-amido-PEG7-acid
Formel:
C₃₂H₄₅NO₁₁ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD26142985 CAS-Nummer: 1863885-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...