Amino- PEG3-t-butyl ester

Lieferant: Apollo Scientific
Achtung

Synonyme: NH2-PEG3-C2-Boc, tert-Butyl 3-(2-(2-(2-aminoethoxy)ethoxy)ethoxy)propanoate

BIPG1072-500MG BIPG1072-250MG BIPG1072-1G
APOSBIPG1072-500MGEA 219 EUR
APOSBIPG1072-500MG APOSBIPG1072-250MG APOSBIPG1072-1G
Amino- PEG3-t-butyl ester
Amino- PEG3-t-butyl ester
Formel: C₁₃H₂₇NO₅
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD06201017
CAS-Nummer: 252881-74-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR