Epibestatin hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,3R)-AHPBA-Leu-OH HCl, ([2R,3R]-3-Amino-2-hydroxy-4-phenylbutanoyl)-L-leucine
BIE9631-10MG
BIE9631-5MG
BIE9631-1MG
APOSBIE9631-10MGEA
432
EUR
APOSBIE9631-10MG
APOSBIE9631-5MG
APOSBIE9631-1MG
Epibestatin hydrochloride
Epibestatin hydrochloride
Diastereoepimer of aminopeptidase inhibitor bestatin
Formel:
C₁₆H₂₄N₂O₄·ClH Molekulargewicht: 344.834 g/mol Lagerungstemperatur: Gefrierschrank |
MDL Number:
MFCD00070386 CAS-Nummer: 100992-60-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...