Methyl 3,4-di-O-acetyl-D-glucuronal ≥99%
Lieferant: Apollo Scientific
Synonyme:
1,5-Anhydro-2-deoxy-D-arabino-hex-5-enonic acid diacetate
BICL4141-1G
BICL4141-500MG
BICL4141-250MG
APOSBICL4141-1GEA
1110
EUR
APOSBICL4141-1G
APOSBICL4141-500MG
APOSBICL4141-250MG
Methyl 3,4-di-O-acetyl-D-glucuronal ≥99%
Methyl 3,4-di-O-acetyl-D-glucuronal
Lagerungstemperatur:
Raumtemperatur |
CAS-Nummer:
57690-62-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...