2,3-O-Isopropylidene-L-gulono-1,4-lactone ≥98%
Lieferant: Apollo Scientific
Synonyme:
2,3-O-Isopropylidene-L-gulonic acid γ-lactone, L-Gulonolactone acetonide
BICL4123-1G
BICL4123-5G
BICL4123-2G
APOSBICL4123-1GEA
447
EUR
APOSBICL4123-1G
APOSBICL4123-5G
APOSBICL4123-2G
2,3-O-Isopropylidene-L-gulono-1,4-lactone ≥98%
2,3-O-Isopropylidene-L-gulono-1,4-lactone
Lagerungstemperatur:
Raumtemperatur |
CAS-Nummer:
94840-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...