Formononetin-7-O-β-D-glucuronide, sodium salt ≥99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Biochanin B-7-O-β-D-glucuronide, 7-Hydroxy-4'-methoxyisoflavone-7-O-β-D-glucuronide
BICL4100-10MG
BICL4100-25MG
BICL4100-5MG
APOSBICL4100-10MGEA
420
EUR
APOSBICL4100-10MG
APOSBICL4100-25MG
APOSBICL4100-5MG
Formononetin-7-O-β-D-glucuronide, sodium salt ≥99%
Formononetin-7-O-β-D-glucuronide, sodium salt
Schmelzpunkt:
247…250 °C Lagerungstemperatur: Gefrierschrank |
CAS-Nummer:
18524-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...