Ethyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥99%

Lieferant: Apollo Scientific

Synonyme: (2R,3R,4S,5R,6S)-2-(Acetoxymethyl)-6-(ethylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate

BICL4095-10G BICL4095-5G
APOSBICL4095-10GEA 288 EUR
APOSBICL4095-10G APOSBICL4095-5G
Ethyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside ≥99%
Ethyl 2,3,4,6-tetra-O-acetyl-1-thio-β-D-glucopyranoside
Formel: C₁₆H₂₄O₉S
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04114310
CAS-Nummer: 52645-73-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR