Clavulanate:Cellulose
Lieferant: Apollo Scientific
|
Gefahr
|
BIC581-10G
BIC581-1G
BIC581-5G
APOSBIC581-10GEA
255
EUR
APOSBIC581-10G
APOSBIC581-1G
APOSBIC581-5G
Clavulanate:Cellulose
Clavulanate:Cellulose
Formel:
C₈H₈NO₅·K Lagerungstemperatur: Kühlschrank |
CAS-Nummer:
61177-45-5 EINECS: 262-640-9 UN-Nummer: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...