1-O-Methyl-β-D-glucuronic acid sodium salt

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl β-D-glucuronate Sodium salt

BIB6106-1G BIB6106-500MG BIB6106-5G
APOSBIB6106-1GEA 212 EUR
APOSBIB6106-1G APOSBIB6106-500MG APOSBIB6106-5G
1-O-Methyl-β-D-glucuronic acid sodium salt
1-O-Methyl-β-D-glucuronic acid sodium salt
Formel: C₇H₁₁O₇·Na
Schmelzpunkt: 270…273 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00083405
CAS-Nummer: 134253-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR