5-Benzyloxyindole-3-acetic acid

Lieferant: Apollo Scientific
Achtung

BIB6002-10G BIB6002-5G BIB6002-1G
APOSBIB6002-10GEA 1060 EUR
APOSBIB6002-10G APOSBIB6002-5G APOSBIB6002-1G
5-Benzyloxyindole-3-acetic acid
5-Benzyloxyindole-3-acetic acid
Formel: C₁₇H₁₅NO₃
Schmelzpunkt: 148…152 °C
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00022750
CAS-Nummer: 4382-53-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR