Glycine 7-amido-4-methylcoumarin hydrobromide salt
Lieferant: Apollo Scientific
Synonyme:
H-Gly-amc hbr, 2-Amino-N-(4-methyl-2-oxo-2H-chromen-7-yl)acetamide hydrobromide, Glycine-AMC.HBr
BIB1440-100MG
BIB1440-50MG
APOSBIB1440-100MGEA
197
EUR
APOSBIB1440-100MG
APOSBIB1440-50MG
Glycine 7-amido-4-methylcoumarin hydrobromide salt
Glycine 7-amido-4-methylcoumarin hydrobromide salt
Formel:
C₁₂H₁₂N₂O₃·BrH Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00058452 CAS-Nummer: 113728-13-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...