Azocarmine G, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Rosinduline, Acid red 101, Basalan Carmine GE
M-Clarity™ quality level = MQ100
Standard stain for microscopy. It may be also be used as a reference standard in the determination of azocarmine G in food products and beverages using high performance liquid chromatography coupled with diode array detector (HPLC-DAD).
- Azocarmine G is a synthetic dye, commonly used in food and beverages to restore their original appearance, in situations where, the color is affected by processing, storage, packaging and distribution
- Azocarmine G is useful for protein determination in acidic medium. It is indicated by the formation of purple-red complex
- Azocarmine G may be used as a reference standard for the determination of the analyte in food and beverages using high-performance liquid chromatography coupled with diode-array detector (HPLC-DAD)
Formel:
C₂₈H₁₇N₃Na₂O₆S₂ Lagerungstemperatur: Room Temperature |
MDL Number:
MFCD00042002 CAS-Nummer: 25641-18-3 |
|
Spezifikation Testergebnisse
Appearance (Form) | Powder |
Solubility (Colour) | Red or Purple |
Solubility (Method) | 1 mg/ml in water |
Uv-Vis Spectrum | E1%=210-350 At 510-524Nm In Water |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...