Quinoline yellow sulphonic acid sodium salt (Acid yellow 3) für die Mikroskopie, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY
Achtung

Synonyme: D and C Yellow No. 10, Quinoline Yellow S, Quinaldine Yellow, Food yellow 13, CI 47005

M-Clarity™ Qualitätsstufe = MQ200

22680-100G-F 22680-25G-F
22680-100G-FEA 308 EUR
22680-100G-F 22680-25G-F
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3) für die Mikroskopie, Sigma-Aldrich®
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)

Quinoline Yellow,for microscopy (Hist.), mixture of mono- and disulfonic acid sodium salt (Acid Yellow 3)

Formel: C₁₈H₁₁NO₂.xNaO₃S
Lagerungstemperatur: Room Temperature
MDL Number: MFCD00080727
CAS-Nummer: 8004-92-0

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Yellow to dark yellow and orange to dark orange
Appearance (form) Powder
Thin layer chromatogr. Corresponds to standard
Solubility (colour) 0.1 g in 10 ml water
Solubility (method) 0.1 g in 10 ml water
Infrared spectrum Conforms to structure
Wavelength (1) (UV) 407 - 417 nm
Extinction (1%/1 cm) (1) (UV) ≥450
Wavelength (2) (UV) 283 - 293 nm
Extinction (1%/1 cm) (2) (UV) ≥350
Solvent (UV) Water


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR