5-DTAF hydrochloride, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
5-[(4,6-Dichlorotriazin-2-yl)amino]fluorescein hydrochloride
D0531-100MG
D0531-1G
D0531-500MG
D0531-100MGEA
133
EUR
D0531-100MG
D0531-1G
D0531-500MG
5-DTAF hydrochloride, Sigma-Aldrich®
5-DTAF hydrochloride
5-([4,6-Dichlorotriazin-2-yl]amino)fluorescein hydrochloride has been used for labeling of yeast cells to study the adhesion of yeast cells to the digestive tract of sea bass (Dicentrarchus labrax) larvae and leopard grouper (Mycteroperca rosacea).
Formel:
C₂₃H₁₂Cl₂N₄O₅·HCl Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00051407 CAS-Nummer: 21811-74-5 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...