Amberlite® IRC-120, H form, ion-exchange resin
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
466710010
466710050
466712500
ACRO466710010EA
70.2
EUR
ACRO466710010
ACRO466710050
ACRO466712500
Amberlite® IRC-120, H form, ion-exchange resin
Amberlite® IRC-120, H form, ion-exchange resin
- Inorganic
- for organic synthesis
Lagerungstemperatur:
Raumtemperatur |
MDL Number:
MFCD00132707 CAS-Nummer: 65098-55-7 |
Spezifikation Testergebnisse
Appearance | Amber to light brown Beads |
Particle size | ≤4.0 % (> 1180um) |
Moisture holding capacity | 48.0 to 58.0 (H+) |
Total volume capacity | ≥1.80 eq/L (H+) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...