Pravastatin sodium salt hydrate
Lieferant: Thermo Fisher Scientific
|
Achtung
|
464880010
464882500
ACRO464880010EA
340
EUR
ACRO464880010
ACRO464882500
Pravastatin sodium salt hydrate
Pravastatin sodium salt hydrate
- Bioactive
Formel:
C₂₃H₃₅O₇Na·xH₂O Schmelzpunkt: 171…173 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00887601 CAS-Nummer: 2244386-63-6 |
Spezifikation Testergebnisse
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Water | ≤4 % |
Specific optical rotation | +150° to +159° (20°C, 589 nm) (0.5% in water) |
Titration with HClO4 | 97.0 to 102.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...