10H-Phenothiazine-10-propanesulphonic acid sodium salt
Lieferant: Thermo Fisher Scientific
Synonyme:
Sodium 3-(10H-phenothiazin-10-yl)propane-1-sulphonate
464000010
464000050
ACRO464000010EA
113
EUR
ACRO464000010
ACRO464000050
10H-Phenothiazine-10-propanesulphonic acid sodium salt
10H-Phenothiazine-10-propanesulphonic acid sodium salt
- Bioactive
Formel:
C₁₅H₁₄NNaO₃S₂ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD08275674 CAS-Nummer: 101199-38-6 |
Spezifikation Testergebnisse
Appearance | Almost white to grey to brown Powder |
Infrared spectrum | Conforms |
Titration | 97.5 to 102.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...