Risedronic acid sodium salt

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Risedronic acid sodium, (1-Hydroxy-2-(pyridin-3-yl)ethane-1,1-diyl)bis(phosphonic acid) sodium salt

462930010 462932500
ACRO462930010EA 263 EUR
ACRO462930010 ACRO462932500
Risedronic acid sodium salt
Risedronic acid sodium salt

  • Bioactive

Formel: C₇H₁₀NNaO₇P₂
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD01706268
CAS-Nummer: 115436-72-1

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Water 11.9 to 13.9 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR