Metoclopramide hydrochloride ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
461820050
461820250
ACRO461820050EA
35.9
EUR
ACRO461820050
ACRO461820250
Metoclopramide hydrochloride ≥98%
Metoclopramide hydrochloride
- Bioactive
Formel:
C₁₄H₂₂ClN₃O₂·HCl Schmelzpunkt: 171…173 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00058011 CAS-Nummer: 7232-21-5 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Water | ≤7.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...