N-Oxalylglycine
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-((Carboxymethyl)amino)-2-oxoacetic acid
464710010
464712500
ACRO464710010EA
765
EUR
ACRO464710010
ACRO464712500
N-Oxalylglycine
N-Oxalylglycine
- Bioactive
Formel:
C₄H₅NO₅ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00913253 CAS-Nummer: 5262-39-5 |
Spezifikation Testergebnisse
Appearance | Colorless to white Solid |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...