Eplerenon
Lieferant: Thermo Fisher Scientific
Synonyme:
9,11alpha-Epoxy-17-hydroxy-3-oxo-17alpha-pregn-4-ene-7alpha,21-dicarboxylic Acid gamma-Lactone Methyl Ester
462970010
462972500
ACRO462970010EA
663
EUR
ACRO462970010
ACRO462972500
Eplerenon
Eplerenone
- Bioactive
Formel:
C₂₄H₃₀O₆ Schmelzpunkt: 242 °C (dec.) Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07783647 CAS-Nummer: 107724-20-9 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤0.5 % |
Sulfated ash | ≤0.1 % |
HPLC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...