6-Propyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinon ≥98%
Lieferant: Thermo Fisher Scientific
|
|
Synonyme:
Propylthiouracil, 4-Hydroxy-2-mercapto-6-propylpyrimidine, 4-Propyl-2-thiouracil, 6-Propyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one
121310250
121311000
ACRO121310250EA
70.4
EUR
ACRO121310250
ACRO121311000
6-Propyl-2-thioxo-2,3-dihydro-4(1H)-pyrimidinon ≥98%
6-Propyl-2-thiouracil
- Bioactive
Formel:
C₇H₁₀N₂OS Schmelzpunkt: 218…221 °C Flammpunkt: 300 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00006041 CAS-Nummer: 51-52-5 |
Spezifikation Testergebnisse
Appearance | White to yellowish white Powder |
Infrared spectrum | Conforms |
Melting point | 217 to 221 °C |
Titration | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...