Rhodium(III)acetylacetonat (≥25,2% Rh) ≥99,99% (Metall-Basis), Premion®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Tris(2,4-pentanedionato-O,O')rhodium (III), Rhodium (III) tris(2,4-pentanedionato-O,O'), Rhodium (III) 2,4-pentanedionate, Rhodium(III) acetylacetonate
10561.02
10561.03
10561.06
10561.02EA
497
EUR
10561.02
10561.03
10561.06
Rhodium(III)acetylacetonat (≥25,2% Rh) ≥99,99% (Metall-Basis), Premion®
Rhodium(III)acetylacetonat
Purity: 99.99%
CAS No.: 14284-92-5
Molecular Formula: C15H21O6Rh
Formula Weight: 400.23
Physical Form: Crystalline
Melting Point: 263-264°
MDL No.: MFCD00083144
Formel:
C₁₅H₂₁O₆Rh Schmelzpunkt: 263…264 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00083144 CAS-Nummer: 14284-92-5 EINECS: 238-192-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...