Iridium(III)acetylacetonat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Iridium(III) 2,4-acetylacetonate, Tris(2,4-pentanedionato-O,O')iridium (III), Iridium (III) tris(2,4-pentanedionato-O,O'), Iridium (III) 2,4-pentanedionate
M-Clarity™ quality level = MQ100
333352-2.5G
333352-500MG
SIAL333352-2.5GEA
1130
EUR
SIAL333352-2.5G
SIAL333352-500MG
Iridium(III)acetylacetonat, Sigma-Aldrich®
Iridium(III)acetylacetonat
Formel:
C₁₅H₂₁IrO₆ |
MDL Number:
MFCD00015353 CAS-Nummer: 15635-87-7 EINECS: 239-711-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...