Iridium(III)acetylacetonat (≥37,5% Ir)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Iridium(III) 2,4-acetylacetonate, Tris(2,4-pentanedionato-O,O')iridium (III), Iridium (III) tris(2,4-pentanedionato-O,O'), Iridium (III) 2,4-pentanedionate
10514.01
10514.03
10514.06
10514.01EA
128
EUR
10514.01
10514.03
10514.06
Iridium(III)acetylacetonat (≥37,5% Ir)
Iridium(III)acetylacetonat
CAS No.: 15635-87-7
Molecular Formula: C15H21IrO6
Formula Weight: 489.53
Physical Form: Crystalline
Appearance: Orange-yellow
Odour: Odourless
MDL No.: MFCD00015353
Formel:
C₁₅H₂₁IrO₆ Schmelzpunkt: 269…271 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00015353 CAS-Nummer: 15635-87-7 EINECS: 239-711-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...