Cobalt(II)carbonat ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Cobaltous carbonate, Cobalt carbonate, Cobalt(II) carbonate

42745.18 42745.30
42745.18EA 95.2 EUR
42745.18 42745.30
Cobalt(II)carbonat ≥99,5% (Metall-Basis)
Cobalt(II) carbonate

Purity: 99.5% CAS No.: 513-79-1 Molecular Formula: CoCO3 Formula Weight: 118.94 Physical Form: -325 Mesh Powder Density: 4.13 Melting Point: dec. Merck Reference: 14,2436 UN No.: 3077 MDL No.: MFCD00010940

Formel: CCoO₃
Schmelzpunkt: Dec.
Dichte: 4.13
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010940
CAS-Nummer: 513-79-1
EINECS: 208-169-4
UN-Nummer: 3077
ADR: 9,III
Merck Index: 14,2436

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR