Molybdän(VI)oxid ≥99,5%, Puriss. p.a. analytical reagent, Fluka™

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Molybdenum trioxide

69850-100G 69850-500G
69850-100GEA 115.41 EUR
69850-100G 69850-500G
Molybdän(VI)oxid ≥99,5%, Puriss. p.a. analytical reagent, Fluka™
Molybdenum(VI) oxide
Formel: MoO₃
Siedepunkt: 1.155 °C
Schmelzpunkt: 795 °C
Dichte: 4.70 g/cm³ (21 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00003469
CAS-Nummer: 1313-27-5
EINECS: 215-204-7
ADR: 6.1,III
Merck Index: 13,06261

Bestellen

Spezifikation Testergebnisse

Assay Min. 99.8 %
Colour of the solution complying
Turbidity of solution Max. 5.5 NTU
Calcium (Ca) Max. 20 ppm
Cadmium (Cd) Max. 10 ppm
Cobalt (Co) Max. 20 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 20 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 10 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 20 ppm
Lead (Pb) Max. 20 ppm
Zinc (Zn) Max. 20 ppm
Chloride (Cl) Max. 20 ppm
Sulfate (SO₄) Max. 50 ppm
Ammonium (NH₄) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR