Molybdän(VI)oxid 99+%

Lieferant: Thermo Fisher Scientific

206361000 206365000
ACRO206361000EA 66.4 EUR
ACRO206361000 ACRO206365000
Molybdän(VI)oxid 99+%

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to green to grey Powder
Assay ≥99.0 %
Chloride (Cl) ≤50 ppm
Sulfate (SO4) ≤200 ppm
PO4, AsO4, SiO4 (as PO4) ≤0.002 %
Ammonium (NH4) ≤100 ppm
Iron (Fe) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR