Molybdäntrichlorid ≥99,5% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
14034.04
14034.09
14034.04EA
105
EUR
14034.04
14034.09
Molybdäntrichlorid ≥99,5% (Metall-Basis)
Molybdäntrichlorid
Purity: 99.5%
CAS No.: 13478-18-7
Molecular Formula: MoCl3
Formula Weight: 202.32
Physical Form: -100 Mesh Powder
Sensitivity: Air and Moisture Sensitive
Appearance: Dark red
Odour: Acidic
Melting Point: dec.
UN No.: 3260
MDL No.: MFCD00014214
Formel:
Cl₃Mo Schmelzpunkt: Dec. Dichte: 3.58 g/cm³ (25 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00014214 CAS-Nummer: 13478-18-7 EINECS: 236-775-9 UN-Nummer: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...