Kupfer(II)carbonat basisch ≥95% (RT), Purum p.a. analytical reagent

Lieferant: Honeywell Chemicals
Achtung

Synonyme: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)

61167-250G 61167-1KG
61167-250GEA 65.81 EUR
61167-250G 61167-1KG
Kupfer(II)carbonat basisch ≥95% (RT), Purum p.a. analytical reagent
Kupfer(II)carbonat basisch

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: CuCO₃·Cu(OH)₂
Schmelzpunkt: 270 °C
Dichte: 4 g/cm³
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00010976
CAS-Nummer: 12069-69-1
EINECS: 235-113-6
ADR: 6.1,III
Merck Index: 13,02658

Bestellen

Spezifikation Testergebnisse

Assay 95.0 - 105.0 %
Calcium (Ca) Max. 5000 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Iron (Fe) Max. 100 ppm
Potassium (K) Max. 100 ppm
Sodium (Na) Max. 5000 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 1000 ppm
Total S (as SO₄) Max. 200 ppm
Chloride (Cl) Max. 1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR