Samarium(II)iodid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Samarium (II) iodide

M-Clarity™ quality level = MQ100

409340-1G 409340-5G
SIAL409340-1GEA 163 EUR
SIAL409340-1G SIAL409340-5G
Samarium(II)iodid, Sigma-Aldrich®
Samarium diiodide
Formel: SmI₂
MDL Number: MFCD00058873
CAS-Nummer: 32248-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR