Ammoniumhydrogensulfat ≥98,0% (durch titrimetrische Analyse), Purum p.a. analytical reagent

Lieferant: Honeywell Chemicals
Gefahr

Synonyme: Ammonium bisulphate

09848-1KG 09848-25KG
09848-1KGEA 93.53 EUR
09848-1KG HONC09848-25KG
Ammoniumhydrogensulfat ≥98,0% (durch titrimetrische Analyse), Purum p.a. analytical reagent
Ammonium hydrogen sulphate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formel: H₅NO₄S
Siedepunkt: 350 °C (1.013 hPa)
Schmelzpunkt: 121-145 °C
Dichte: 1.790 g/cm³
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00051073
CAS-Nummer: 7803-63-6
EINECS: 232-265-5
UN-Nummer: 2506
ADR: 8,II
Merck Index: 12,00526

Bestellen

Spezifikation Testergebnisse

Assay 98.0 - 102.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 500 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR