Tetrabutylammoniumhexafluorophosphat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetrabutylammonium hexafluorophosphate(V), Tetra-n-butylammonium hexafluorophosphate
M-Clarity™ quality level = MQ100
281026-5G
281026-100G
281026-25G
SIAL281026-5GEA
32.9
EUR
SIAL281026-5G
SIAL281026-100G
SIAL281026-25G
Tetrabutylammoniumhexafluorophosphat, Sigma-Aldrich®
Tetrabutylammonium hexafluorophosphate
Formel:
C₁₆H₃₆F₆NP |
MDL Number:
MFCD00011748 CAS-Nummer: 3109-63-5 EINECS: 221-472-6 UN-Nummer: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...