Tetrabutylammoniumcyanid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ quality level = MQ200
358665-1G
358665-5G
SIAL358665-1GEA
62.3
EUR
SIAL358665-1G
SIAL358665-5G
Tetrabutylammoniumcyanid, Sigma-Aldrich®
Tetrabutylammonium cyanide
Formel:
C₁₇H₃₆N₂ Lagerungstemperatur: Raumtemperatur |
CAS-Nummer:
10442-39-4 EINECS: 233-926-0 UN-Nummer: 2811 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...