Tetrabutylammoniumperchlorat elektrochemischer Grad
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Tetra-n-butylammonium perchlorate
CAS No.: 1923-70-2
Molecular Formula: C16H36ClNO4
Formula Weight: 341.92
Physical Form: Crystalline
Sensitivity: Hygroscopic
Appearance: White
Melting Point: 210-217°
Solubility: Soluble in ethanol. Very slightly soluble in water
UN No.: 1481
MDL No.: MFCD00038722
Suitable for use as a polarographic supporting electrolyte
Formel:
C₁₆H₃₆ClNO₄ Molekulargewicht: 341.92 g/mol Schmelzpunkt: 210…217 °C Dichte: 1.07 g/cm³ (20 °C) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00038722 CAS-Nummer: 1923-70-2 EINECS: 217-655-5 UN-Nummer: 1481 ADR: 5.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...