Dodecyltrimethylammoniumbromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide

M-Clarity™ Qualitätsstufe = MQ200

D8638-100G D8638-25G
SIALD8638-100GEA 597 EUR
SIALD8638-100G SIALD8638-25G
Dodecyltrimethylammoniumbromid, Sigma-Aldrich®
Dodecyltrimethylammonium bromide
Formel: C₁₅H₃₄BrN
Molekulargewicht: 308.35 g/mol
Schmelzpunkt: 264…266 °C
MDL Number: MFCD00011767
CAS-Nummer: 1119-94-4
EINECS: 214-290-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR