Bleititanat ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Lead titanium oxide, Lead(II) titanium oxide, Lead(II) titanate, Lead titanate (IV), Lead titanate
35671.18
35671.30
35671.18EA
103
EUR
35671.18
35671.30
Bleititanat ≥99,9% (Metall-Basis)
Bleititanat
Purity: 99.9%
CAS No.: 12060-00-3
Molecular Formula: PbTiO3
Formula Weight: 303.09
Physical Form: -200 Mesh Powder
Density: 7.520
Appearance: Yellow
Odour: Odourless
UN No.: 3288
MDL No.: MFCD00011159
Formel:
O₃PbTi Schmelzpunkt: 1120 °C Dichte: 7.520 Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00011159 CAS-Nummer: 12060-00-3 EINECS: 235-038-9 UN-Nummer: 3288 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...